Assay Details
Target Gene Details
Entrez Gene ID: | 6487 |
Gene Name: | ST3 beta-galactoside alpha-2,3-sialyltransferase 3 |
Gene Aliases: |
EIEE15, MRT12, SIAT6, ST3GALII, ST3GalIII, ST3N |
Location: |
Chr.1:43707497-43931165 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
More Information
Set Membership: |
|

Genomic Map