Assay Details
Target Gene Details
Entrez Gene ID: | 55192 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C17 |
Gene Aliases: |
- |
Location: |
Chr.15:40767448-40807478 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC17 | NM_018163.2 | NP_060633.1 | ||
| XM_017022401.1 | XP_016877890.1 | |||
| XM_017022402.1 | XP_016877891.1 | |||
| AK001496.1 | BAA91724.1 | |||
| AK308659.1 | ||||
| AK308858.1 | ||||
| AK309307.1 | ||||
| AK310140.1 | ||||
| BC000048.2 | AAH00048.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73398528] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map