Assay Details
Target Gene Details
Entrez Gene ID: | 9616 |
Gene Name: | ring finger protein 7 |
Gene Aliases: |
CKBBP1, ROC2, SAG |
Location: |
Chr.3:141738209-141746803 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF7 | NM_001201370.1 | NP_001188299.1 | ||
| NM_014245.4 | NP_055060.1 | |||
| NM_183237.2 | NP_899060.1 | |||
| NR_037702.1 | ||||
| NR_037703.1 | ||||
| AF092878.1 | AAD25962.1 | |||
| AF142060.1 | AAD30147.1 | |||
| AF164679.1 | AAD55984.1 | |||
| AF312226.1 | AAK37450.1 | |||
| AK289894.1 | ||||
| AK311982.1 | ||||
| BC005966.1 | AAH05966.1 | |||
| BC008627.1 | AAH08627.1 | |||
| BG715867.1 | ||||
| BG716023.1 | ||||
| BG720797.1 | ||||
| BG742338.1 | ||||
| BT007348.1 | AAP36012.1 | |||
| DB272382.1 |
More Information
Set Membership: |
|

Genomic Map