Assay Details
Target Gene Details
Entrez Gene ID: | 27133 |
Gene Name: | potassium voltage-gated channel subfamily H member 5 |
Gene Aliases: |
EAG2, H-EAG2, Kv10.2, hEAG2 |
Location: |
Chr.14:62706573-63045238 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNH5 | NM_139318.4 | 11 | 2855 | NP_647479.2 |
| NM_172375.2 | 10 | 2658 | NP_758963.1 | |
| AF418206.1 | 11 | 2803 | AAM28435.1 | |
| AF472412.1 | 11 | 2803 | AAM49565.1 | |
| AF493798.1 | 10 | 2606 | AAM49574.1 | |
| BC073979.1 | 11 | 2855 | AAH73979.1 | |
| U69185.1 | 3 | 1245 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527675 | Chr.14:62701829 - 62744562 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map