Assay Details
Target Gene Details
Entrez Gene ID: | 8061 |
Gene Name: | FOS like 1, AP-1 transcription factor subunit |
Gene Aliases: |
FRA, FRA1, fra-1 |
Location: |
Chr.11:65892136-65900526 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOSL1 | NM_001300844.1 | NP_001287773.1 | ||
| NM_001300855.1 | NP_001287784.1 | |||
| NM_001300856.1 | NP_001287785.1 | |||
| NM_001300857.1 | NP_001287786.1 | |||
| NM_005438.4 | NP_005429.1 | |||
| NR_125339.1 | ||||
| AK222902.1 | BAD96622.1 | |||
| AK293297.1 | ||||
| AK299050.1 | ||||
| AK313718.1 | ||||
| BC016648.1 | AAH16648.1 | |||
| BM764259.1 | ||||
| BM846271.1 | ||||
| BP222547.1 | ||||
| BQ937004.1 | ||||
| CB123614.1 | ||||
| CR542257.1 | CAG47053.1 | |||
| CR542278.1 | CAG47074.1 | |||
| KU178407.1 | ||||
| KU178408.1 | ||||
| X16707.1 | CAA34679.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759834 | Chr.11:65792422 - 65954892 on Build GRCh38 | Gain |
|
| nsv832191 | Chr.11:65864858 - 66038649 on Build GRCh38 | Loss |
|
| nsv951019 | Chr.11:65874630 - 65899129 on Build GRCh38 | Deletion |
|
| nsv555215 | Chr.11:65814108 - 65909045 on Build GRCh38 | Loss |
|
| nsv1159905 | Chr.11:65836608 - 65922038 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map