Assay Details
Target Gene Details
Entrez Gene ID: | 2002 |
Gene Name: | ELK1, ETS transcription factor |
Gene Aliases: |
- |
Location: |
Chr.X:47635520-47650604 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELK1 | NM_001114123.2 | 5 | 1044 | NP_001107595.1 |
| NM_001257168.1 | NP_001244097.1 | |||
| NM_005229.4 | 4 | 938 | NP_005220.2 | |
| XM_017029339.1 | 3 | 759 | XP_016884828.1 | |
| AB016193.1 | 4 | 822 | BAA36616.1 | |
| AB209047.1 | 3 | 2804 | BAD92284.1 | |
| AF000672.1 | AAD00862.1 | |||
| AK093966.1 | BAC04259.1 | |||
| AK312984.1 | 5 | 899 | ||
| BC048296.1 | 4 | 800 | AAH48296.1 | |
| BC056150.1 | 3 | 768 | AAH56150.1 | |
| CR542252.1 | 3 | 722 | CAG47048.1 | |
| M25269.1 | 5 | 1037 | AAA52384.1 |
More Information
Set Membership: |
|

Genomic Map