Assay Details
Target Gene Details
Entrez Gene ID: | 163479 |
Gene Name: | fibronectin type III domain containing 7 |
Gene Aliases: |
- |
Location: |
Chr.1:108712934-108742749 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FNDC7 | NM_001144937.1 | 9 | 1686 | NP_001138409.1 |
| AK093157.1 | 5 | 1012 | BAC04077.1 | |
| BC057762.1 | 5 | 1059 | AAH57762.1 | |
| BC130421.1 | 5 | 1012 |
More Information
Additional Information:
For this assay, SNP(s) [rs76462144] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map