Assay Details
Target Gene Details
Entrez Gene ID: | 140456 |
Gene Name: | ankyrin repeat and SOCS box containing 11 |
Gene Aliases: |
- |
Location: |
Chr.X:15281697-15315624 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB11 | NM_001012428.2 | 5 | 597 | NP_001012428.1 |
| NM_001201583.1 | 5 | 555 | NP_001188512.1 | |
| NM_080873.2 | 5 | 606 | NP_543149.1 | |
| AF425642.1 | 5 | 587 | AAL60519.1 | |
| BC069340.1 | 5 | 587 | AAH69340.1 | |
| BC103874.1 | 5 | 587 | AAI03875.1 | |
| BC103875.1 | 5 | 587 | AAI03876.1 | |
| BX537853.1 | 5 | 597 | CAD97861.1 | |
| BX537857.1 | 5 | 555 | CAD97864.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522729 | Chr.X:15283411 - 15334052 on Build GRCh38 | Gain |
|
| nsv528343 | Chr.X:15271931 - 15385229 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map