Assay Details
Target Gene Details
Entrez Gene ID: | 161291 |
Gene Name: | transmembrane protein 30B |
Gene Aliases: |
CDC50B |
Location: |
Chr.14:61277370-61281812 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM30B | NM_001017970.2 | 1 | 3886 | NP_001017970.1 |
| AK091169.1 | 2 | 2787 | ||
| AL080078.1 | 1 | 1646 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564881 | Chr.14:60457291 - 61377535 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map