Assay Details
Target Gene Details
Entrez Gene ID: | 127294 |
Gene Name: | myomesin 3 |
Gene Aliases: |
- |
Location: |
Chr.1:24056041-24112175 on Build GRCh38 |
Assay Gene Location: | Within Exon 37 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYOM3 | NM_152372.3 | 37 | 5509 | NP_689585.3 |
| AK093280.1 | 10 | 2395 | BAC04119.1 | |
| AK128233.1 | 29 | 4569 | BAC87343.1 | |
| BC014671.1 | 11 | 2029 | AAH14671.1 |
More Information
Set Membership: |
|

Genomic Map