Assay Details
Target Gene Details
Entrez Gene ID: | 388818 |
Gene Name: | keratin associated protein 26-1 |
Gene Aliases: |
- |
Location: |
Chr.21:30319131-30320289 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTAP26-1 | NM_203405.1 | 1 | 64 | NP_981950.1 |
| BC057825.1 | 1 | 65 | AAH57825.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834076 | Chr.21:30216502 - 30383428 on Build GRCh38 | Gain |
|
| nsv1057467 | Chr.21:30318003 - 30993921 on Build GRCh38 | Gain |
|
| nsv1059954 | Chr.21:30301334 - 30339600 on Build GRCh38 | Loss |
|
| nsv587354 | Chr.21:29978351 - 30620545 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78731676] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map