Assay Details
Target Gene Details
Entrez Gene ID: | 83657 |
Gene Name: | dynein light chain roadblock-type 2 |
Gene Aliases: |
DNCL2B, DNLC2B, ROBLD2 |
Location: |
Chr.16:80540259-80550811 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DYNLRB2 | NM_001305017.1 | 3 | 266 | NP_001291946.1 |
| NM_130897.2 | 3 | 273 | NP_570967.1 | |
| NR_130942.1 | 3 | 814 | ||
| NR_130943.1 | 4 | 363 | ||
| XM_006721295.3 | 3 | 470 | XP_006721358.1 | |
| XM_006721296.3 | 3 | 223 | XP_006721359.1 | |
| XM_011523370.2 | 3 | 230 | XP_011521672.1 | |
| AF125108.1 | 3 | 268 | AAL75945.1 | |
| BC054892.1 | 4 | 294 | AAH54892.1 | |
| CD676360.1 | 3 | 266 | ||
| DB458006.1 | 3 | 235 | ||
| HY018370.1 | 3 | 274 | ||
| HY123054.1 | 3 | 256 | ||
| HY201608.1 | 2 | 395 |
Target Gene Details
Entrez Gene ID: | 102724084 |
Gene Name: | uncharacterized LOC102724084 |
Gene Aliases: |
- |
Location: |
Chr.16:80155958-80563135 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724084 | NR_120307.1 | |||
| DA228026.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833300 | Chr.16:80441169 - 80567139 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map