Assay Details
Target Gene Details
Entrez Gene ID: | 387032 |
Gene Name: | zinc finger with KRAB and SCAN domains 4 |
Gene Aliases: |
P1P373C6, ZNF307, ZNF427, ZSCAN36 |
Location: |
Chr.6:28241697-28259312 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZKSCAN4 | NM_001304506.1 | NP_001291435.1 | ||
| NM_019110.4 | NP_061983.2 | |||
| XM_005249095.3 | XP_005249152.1 | |||
| XM_005249097.3 | XP_005249154.1 | |||
| XM_005249098.3 | XP_005249155.1 | |||
| XM_005249100.4 | XP_005249157.2 | |||
| XM_006715095.3 | XP_006715158.1 | |||
| XM_011514588.2 | XP_011512890.1 | |||
| XM_011514589.2 | XP_011512891.1 | |||
| XM_017010844.1 | XP_016866333.1 | |||
| AK302017.1 | ||||
| AK315783.1 | ||||
| AY781778.1 | AAV41023.1 | |||
| BC014031.2 | AAH14031.1 | |||
| KU178905.1 | ||||
| KU178906.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830620 | Chr.6:28206219 - 28380256 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map