Assay Details
Target Gene Details
Entrez Gene ID: | 3169 |
Gene Name: | forkhead box A1 |
Gene Aliases: |
HNF3A, TCF3A |
Location: |
Chr.14:37589552-37595120 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXA1 | NM_004496.3 | 2 | 1991 | NP_004487.2 |
| XM_017021246.1 | 2 | 2553 | XP_016876735.1 | |
| BC033890.1 | 2 | 1905 | AAH33890.1 | |
| U39840.1 | 2 | 1930 | AAB06493.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832772 | Chr.14:37561095 - 37756716 on Build GRCh38 | Loss |
|
| nsv826923 | Chr.14:37548414 - 37639440 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map