Assay Details
Target Gene Details
Entrez Gene ID: | 56521 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C12 |
Gene Aliases: |
JDP1 |
Location: |
Chr.10:67796665-67838179 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC12 | NM_021800.2 | NP_068572.1 | ||
| NM_201262.1 | 3 | 499 | NP_957714.1 | |
| XM_017016431.1 | XP_016871920.1 | |||
| XM_017016432.1 | XP_016871921.1 | |||
| AF176012.1 | AAD52650.1 | |||
| AF176013.1 | 3 | 416 | AAD52651.1 | |
| AK289537.1 | ||||
| BC017018.1 | AAH17018.1 | |||
| CB121724.1 | ||||
| CR457184.1 | CAG33465.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551433 | Chr.10:67805187 - 67937238 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map