Assay Details
Target Gene Details
Entrez Gene ID: | 80777 |
Gene Name: | cytochrome b5 type B |
Gene Aliases: |
CYB5-M, CYPB5M, OMB5 |
Location: |
Chr.16:69424595-69466264 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYB5B | NM_030579.2 | 5 | 3634 | NP_085056.2 |
| AK098148.1 | 1 | 1769 | ||
| AK125218.1 | 1 | 1172 |
More Information
Additional Information:
For this assay, SNP(s) [rs111281462] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map