Assay Details
Target Gene Details
Entrez Gene ID: | 1738 |
Gene Name: | dihydrolipoamide dehydrogenase |
Gene Aliases: |
DLDD, DLDH, E3, GCSL, LAD, PHE3 |
Location: |
Chr.7:107891107-107921198 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLD | NM_000108.4 | NP_000099.2 | ||
| NM_001289750.1 | NP_001276679.1 | |||
| NM_001289751.1 | NP_001276680.1 | |||
| NM_001289752.1 | NP_001276681.1 | |||
| AB209703.1 | BAD92940.1 | |||
| AK225289.1 | ||||
| AK225301.1 | ||||
| AK294146.1 | ||||
| AK295080.1 | ||||
| AK297508.1 | ||||
| AK300077.1 | ||||
| AK310220.1 | ||||
| AK312346.1 | ||||
| BC018648.2 | AAH18648.1 | |||
| BC018696.2 | AAH18696.1 | |||
| J03490.1 | AAA59527.1 | |||
| J03620.1 | AAA35764.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv949927 | Chr.7:107890656 - 107915255 on Build GRCh38 | Duplication |
|
| nsv831088 | Chr.7:107856655 - 108053235 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75123588] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map