Assay Details
Target Gene Details
Entrez Gene ID: | 55284 |
Gene Name: | ubiquitin conjugating enzyme E2 W (putative) |
Gene Aliases: |
UBC-16, UBC16 |
Location: |
Chr.8:73780097-73878910 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2W | NM_001001481.2 | NP_001001481.2 | ||
| NM_001271015.1 | NP_001257944.1 | |||
| NM_018299.4 | NP_060769.4 | |||
| NR_073119.1 | ||||
| NR_073120.1 | ||||
| NR_073121.1 | ||||
| AK001873.1 | BAA91954.1 | |||
| AK021735.1 | BAB13883.1 | |||
| AK024050.1 | BAB14800.1 | |||
| AK225554.1 | ||||
| AK295792.1 | ||||
| AY948289.1 | AAY24555.1 | |||
| BC010900.1 | AAH10900.1 | |||
| BC016326.1 | AAH16326.1 | |||
| BU155172.1 | ||||
| BX538145.1 | ||||
| BX648427.1 | ||||
| DA352184.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs144174398] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map