Assay Details
Target Gene Details
Entrez Gene ID: | 146760 |
Gene Name: | reticulon 4 receptor like 1 |
Gene Aliases: |
NGRH2, NgR3 |
Location: |
Chr.17:1934677-2025345 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTN4RL1 | NM_178568.3 | 2 | 2804 | NP_848663.1 |
| AL834409.1 | 1 | 2270 | CAD39071.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33335 | Chr.17:1779713 - 2048672 on Build GRCh38 | Loss |
|
| nsv1056771 | Chr.17:1873573 - 1947080 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map