Assay Details
Target Gene Details
Entrez Gene ID: | 7482 |
Gene Name: | Wnt family member 2B |
Gene Aliases: |
WNT13 |
Location: |
Chr.1:112466541-112521288 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WNT2B | NM_001291880.1 | 3 | 1427 | NP_001278809.1 |
| NM_004185.4 | 4 | 1498 | NP_004176.2 | |
| NM_024494.2 | 3 | 1073 | NP_078613.1 | |
| AB045116.1 | 4 | 621 | BAB11984.1 | |
| AB045117.1 | 3 | 577 | BAB11985.1 | |
| AK127449.1 | 3 | 1427 | ||
| AK312696.1 | 3 | 825 | ||
| BC141825.1 | 3 | 1073 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831081 | Chr.1:112392298 - 112580018 on Build GRCh38 | Loss |
|
| esv27061 | Chr.1:112150007 - 112703641 on Build GRCh38 | Gain+Loss |
|
| nsv1008375 | Chr.1:112485649 - 112517599 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map