Assay Details
Target Gene Details
Entrez Gene ID: | 27033 |
Gene Name: | zinc finger and BTB domain containing 32 |
Gene Aliases: |
FAXF, FAZF, Rog, TZFP, ZNF538 |
Location: |
Chr.19:35704481-35717048 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB32 | NM_001316902.1 | 7 | 1092 | NP_001303831.1 |
| NM_001316903.1 | 6 | 1018 | NP_001303832.1 | |
| NM_014383.2 | 7 | 2032 | NP_055198.1 | |
| XM_011526718.2 | 6 | 2193 | XP_011525020.1 | |
| XM_017026588.1 | 4 | 1806 | XP_016882077.1 | |
| XM_017026589.1 | 7 | 2403 | XP_016882078.1 | |
| XM_017026590.1 | 3 | 1804 | XP_016882079.1 | |
| XM_017026591.1 | 6 | 1206 | XP_016882080.1 | |
| XM_017026592.1 | 4 | 886 | XP_016882081.1 | |
| AF130255.1 | 6 | 1884 | AAD27708.1 | |
| BC017700.2 | 4 | 1657 | AAH17700.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv579422 | Chr.19:35665686 - 35774606 on Build GRCh38 | Loss |
|
| nsv828544 | Chr.19:35716289 - 35719856 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73048513] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map