Assay Details
Target Gene Details
Entrez Gene ID: | 9813 |
Gene Name: | EF-hand calcium binding domain 14 |
Gene Aliases: |
KIAA0494 |
Location: |
Chr.1:46675159-46719064 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB14 | NM_014774.2 | NP_055589.1 | ||
| AB007963.1 | ||||
| AK001487.1 | ||||
| AK295722.1 | ||||
| BC002525.2 | AAH02525.1 |
Target Gene Details
Entrez Gene ID: | 100130197 |
Gene Name: | EFCAB14 antisense RNA 1 |
Gene Aliases: |
KIAA0494-AS1 |
Location: |
Chr.1:46674036-46692098 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB14-AS1 | NR_038827.1 | |||
| NR_038828.1 | ||||
| AA993124.1 | ||||
| BE504902.1 | ||||
| BX113937.1 |
More Information
Set Membership: |
|

Genomic Map