Assay Details
Target Gene Details
Entrez Gene ID: | 159296 |
Gene Name: | NK2 homeobox 3 |
Gene Aliases: |
CSX3, NK2.3, NKX2.3, NKX2C, NKX4-3 |
Location: |
Chr.10:99532933-99536524 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKX2-3 | NM_145285.2 | 2 | 1231 | NP_660328.2 |
| AK300856.1 | 2 | 1183 | ||
| AK313485.1 | ||||
| BC025788.1 | 3 | 1114 | AAH25788.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831958 | Chr.10:99451304 - 99651424 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map