Assay Details
Target Gene Details
Entrez Gene ID: | 950 |
Gene Name: | scavenger receptor class B member 2 |
Gene Aliases: |
AMRF, CD36L2, EPM4, HLGP85, LGP85, LIMP-2, LIMPII, SR-BII |
Location: |
Chr.4:76158737-76213899 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCARB2 | NM_001204255.1 | 9 | 2697 | NP_001191184.1 |
| NM_005506.3 | 12 | 3126 | NP_005497.1 | |
| AK056001.1 | 1 | 369 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470047 | Chr.4:76112437 - 76175624 on Build GRCh38 | Gain |
|
| nsv1000178 | Chr.4:76131893 - 76191927 on Build GRCh38 | Gain |
|
| nsv461558 | Chr.4:76112572 - 76194400 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map