Assay Details
Target Gene Details
Entrez Gene ID: | 26263 |
Gene Name: | F-box protein 22 |
Gene Aliases: |
FBX22, FISTC1 |
Location: |
Chr.15:75903859-75937513 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXO22 | NM_012170.3 | 6 | 1569 | NP_036302.1 |
| NM_147188.2 | NP_671717.1 | |||
| NR_037623.1 | ||||
| XM_011521447.2 | XP_011519749.1 | |||
| AK223500.1 | BAD97220.1 | |||
| AK294311.1 | ||||
| AK307084.1 | 4 | 1368 | ||
| AL705285.1 | 1 | 379 | ||
| AL705308.1 | 1 | 379 | ||
| AY005144.1 | AAF89095.1 | |||
| BC020204.1 | ||||
| BC032540.1 | AAH32540.1 | |||
| BC039024.1 | AAH39024.1 | |||
| BC041691.1 | AAH41691.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2749895 | Chr.15:75415580 - 76472296 on Build GRCh38 | Deletion |
|
| nsv1044733 | Chr.15:75828583 - 75936539 on Build GRCh38 | Loss |
|
| esv3636933 | Chr.15:75908231 - 76025938 on Build GRCh38 | Gain |
|
| esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
|
| dgv2622n100 | Chr.15:75904333 - 76022201 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map