Assay Details
Target Gene Details
Entrez Gene ID: | 338692 |
Gene Name: | ankyrin repeat domain 13D |
Gene Aliases: |
- |
Location: |
Chr.11:67289291-67302484 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD13D | NM_207354.2 | NP_997237.2 | ||
| NR_030767.1 | ||||
| XM_011544985.1 | XP_011543287.1 | |||
| XM_011544986.1 | XP_011543288.1 | |||
| XM_011544987.2 | XP_011543289.1 | |||
| XM_011544988.2 | XP_011543290.1 | |||
| AK027313.1 | ||||
| AK057566.1 | ||||
| AK057681.1 | ||||
| AK091614.1 | ||||
| AK096277.1 | ||||
| AK124721.1 | BAC85932.1 | |||
| AK126438.1 | BAC86550.1 | |||
| BC044239.1 | 1 | 215 | AAH44239.1 | |
| BC110419.1 | ||||
| BC121024.1 | ||||
| BC121025.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555253 | Chr.11:67265605 - 67397024 on Build GRCh38 | Loss |
|

Genomic Map