Assay Details
Target Gene Details
Entrez Gene ID: | 223 |
Gene Name: | aldehyde dehydrogenase 9 family member A1 |
Gene Aliases: |
ALDH4, ALDH7, ALDH9, E3, TMABADH |
Location: |
Chr.1:165662212-165698663 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH9A1 | NM_000696.3 | 11 | 2355 | NP_000687.3 |
| XM_011509294.2 | 11 | 2419 | XP_011507596.1 | |
| U34252.1 | 11 | 2552 | AAB18827.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs141037630] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map