Assay Details
Target Gene Details
Entrez Gene ID: | 79577 |
Gene Name: | cell division cycle 73 |
Gene Aliases: |
C1orf28, FIHP, HPTJT, HRPT1, HRPT2, HYX |
Location: |
Chr.1:193121958-193254815 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC73 | NM_024529.4 | 2 | 448 | NP_078805.3 |
| XM_006711537.3 | 2 | 372 | XP_006711600.1 | |
| AF312865.1 | 2 | 213 | AAG45339.1 | |
| AK026969.1 | 2 | 340 | BAB15608.1 | |
| AK226038.1 | 2 | 345 | ||
| AK300929.1 | 2 | 307 | ||
| AK314772.1 | 2 | 206 | ||
| BC007325.2 | 2 | 231 | AAH07325.1 | |
| BC065037.1 | 2 | 285 | AAH65037.1 | |
| BM471846.1 | 2 | 401 | ||
| DC298135.1 | 2 | 448 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv466994 | Chr.1:192394185 - 193552205 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map