Assay Details
Target Gene Details
Entrez Gene ID: | 147463 |
Gene Name: | ankyrin repeat domain 29 |
Gene Aliases: |
- |
Location: |
Chr.18:23598926-23662914 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD29 | NM_001308238.1 | 9 | 1655 | NP_001295167.1 |
| NM_173505.3 | 10 | 1754 | NP_775776.2 | |
| XM_011525830.2 | 9 | 1674 | XP_011524132.1 | |
| XM_011525831.2 | 9 | 3694 | XP_011524133.1 | |
| XM_011525832.2 | 9 | 3812 | XP_011524134.1 | |
| XM_017025561.1 | 8 | 1578 | XP_016881050.1 | |
| AK057782.1 | 9 | 1585 | BAB71569.1 | |
| AK123197.1 | 7 | 3074 | BAC85555.1 | |
| BC030622.1 | 10 | 1759 | AAH30622.1 | |
| BM680506.1 | 1 | 536 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34149 | Chr.18:23278175 - 23641927 on Build GRCh38 | Loss |
|
| nsv509688 | Chr.18:23483859 - 23610653 on Build GRCh38 | Insertion |
|

Genomic Map