Assay Details
Target Gene Details
Entrez Gene ID: | 54870 |
Gene Name: | glutamine rich 1 |
Gene Aliases: |
- |
Location: |
Chr.3:49029707-49094373 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| QRICH1 | NM_001320580.1 | 11 | 3386 | NP_001307509.1 |
| NM_001320581.1 | 11 | 3405 | NP_001307510.1 | |
| NM_001320582.1 | 10 | 3345 | NP_001307511.1 | |
| NM_001320583.1 | 10 | 3326 | NP_001307512.1 | |
| NM_001320584.1 | 10 | 2985 | NP_001307513.1 | |
| NM_001320585.1 | 10 | 3185 | NP_001307514.1 | |
| NM_017730.3 | 11 | 3470 | NP_060200.2 | |
| NM_198880.2 | 10 | 3410 | NP_942581.1 | |
| XM_011533863.1 | 10 | 3126 | XP_011532165.1 | |
| AK000266.1 | 8 | 1871 | BAA91042.1 | |
| AK022524.1 | 5 | 1197 | BAB14076.1 | |
| AK025013.1 | 1 | 289 | ||
| AK074313.1 | BAB85047.1 | |||
| AK124205.1 | ||||
| BC000978.2 | 9 | 2844 | AAH00978.2 | |
| BC098375.1 | 8 | 2190 | AAH98375.1 | |
| BC110855.1 | 10 | 3088 | AAI10856.1 |
More Information
Set Membership: |
|

Genomic Map