Assay Details
Target Gene Details
Entrez Gene ID: | 338662 |
Gene Name: | olfactory receptor family 8 subfamily D member 4 |
Gene Aliases: |
OR11-275 |
Location: |
Chr.11:123906432-123907376 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR8D4 | NM_001005197.1 | 1 | 759 | NP_001005197.1 |
| BC137180.1 | 1 | 800 | ||
| BC137206.1 | 1 | 800 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv82n21 | Chr.11:123901052 - 124289279 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map