Assay Details
Target Gene Details
Entrez Gene ID: | 10131 |
Gene Name: | TNF receptor associated protein 1 |
Gene Aliases: |
HSP 75, HSP75, HSP90L, TRAP-1 |
Location: |
Chr.16:3658037-3717597 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAP1 | NM_001272049.1 | 8 | 904 | NP_001258978.1 |
| NM_016292.2 | 9 | 1063 | NP_057376.2 | |
| XM_011522345.1 | 9 | 955 | XP_011520647.1 | |
| XM_017022851.1 | 8 | 989 | XP_016878340.1 | |
| AB209805.1 | 9 | 973 | BAD93042.1 | |
| AF043254.1 | 9 | 965 | AAC02679.1 | |
| AF154108.1 | 9 | 978 | AAF15314.1 | |
| AJ890085.1 | 4 | 1077 | CAI64498.1 | |
| AK057620.1 | 4 | 1077 | ||
| AK093344.1 | 9 | 1016 | BAC04139.1 | |
| AK223049.1 | 9 | 983 | BAD96769.1 | |
| AK223076.1 | 9 | 983 | BAD96796.1 | |
| AK223205.1 | 9 | 983 | BAD96925.1 | |
| AK299127.1 | 8 | 820 | ||
| BC001455.2 | 9 | 961 | AAH01455.2 | |
| BC018950.2 | 9 | 981 | AAH18950.1 | |
| BC023585.2 | 9 | 985 | AAH23585.1 | |
| JX481354.1 | 8 | 891 | ||
| U12595.1 | 9 | 931 | AAA87704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833128 | Chr.16:3525122 - 3690497 on Build GRCh38 | Loss |
|
| nsv518509 | Chr.16:3583656 - 3688960 on Build GRCh38 | Gain |
|
| nsv1051657 | Chr.16:3644759 - 3788297 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map