Assay Details
Target Gene Details
Entrez Gene ID: | 79005 |
Gene Name: | sodium channel modifier 1 |
Gene Aliases: |
- |
Location: |
Chr.1:151166022-151170297 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCNM1 | NM_001204856.1 | 6 | 673 | NP_001191785.1 |
| NM_024041.3 | 6 | 624 | NP_076946.1 | |
| NR_037937.1 | 6 | 574 | ||
| AA374943.1 | 2 | 255 | ||
| AK056322.1 | 6 | 593 | BAB71149.1 | |
| AV718162.2 | 6 | 550 | ||
| BC000264.1 | 6 | 568 | AAH00264.1 | |
| BU540926.1 | 6 | 440 |
Target Gene Details
Entrez Gene ID: | 100534012 |
Gene Name: | TNFAIP8L2-SCNM1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.1:151156629-151170297 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFAIP8L2-SCNM1 | NM_001204848.1 | 6 | 536 | NP_001191777.1 |
| BG531092.1 | 6 | 502 | ||
| CR999387.1 | 5 | 517 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
| esv3587520 | Chr.1:151167937 - 151168786 on Build GRCh38 | Loss |
|
| dgv2n68 | Chr.1:151136983 - 151309302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map