Assay Details
Target Gene Details
Entrez Gene ID: | 84516 |
Gene Name: | dynactin subunit 5 |
Gene Aliases: |
- |
Location: |
Chr.16:23641366-23676207 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCTN5 | NM_001199743.1 | 6 | 1066 | NP_001186672.1 |
| NM_032486.3 | 6 | 1036 | NP_115875.1 | |
| NR_037573.1 | 7 | 1137 | ||
| AK027387.1 | 6 | 899 | BAB55077.1 | |
| AK126469.1 | 1 | 987 | ||
| AK292843.1 | ||||
| AK299631.1 | 6 | 845 | ||
| BC004191.2 | 6 | 916 | AAH04191.1 |
More Information
Set Membership: |
|

Genomic Map