Assay Details
Target Gene Details
Entrez Gene ID: | 57168 |
Gene Name: | aspartate beta-hydroxylase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.22:26429314-26445012 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASPHD2 | NM_020437.4 | 4 | 1603 | NP_065170.2 |
| AK097157.1 | 3 | 948 | ||
| AL161993.1 | 3 | 659 | CAB82325.1 | |
| BC036753.1 | 4 | 1581 | AAH36753.1 |
More Information
Set Membership: |
|

Genomic Map