Assay Details
Target Gene Details
Entrez Gene ID: | 23552 |
Gene Name: | cyclin dependent kinase 20 |
Gene Aliases: |
CCRK, CDCH, P42, PNQALRE |
Location: |
Chr.9:87966441-87974780 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK20 | NM_001039803.2 | 8 | 1588 | NP_001034892.1 |
| NM_001170639.1 | 7 | 1497 | NP_001164110.1 | |
| NM_001170640.1 | 6 | 1401 | NP_001164111.1 | |
| NM_012119.4 | 7 | 1525 | NP_036251.2 | |
| NM_178432.3 | 7 | 1564 | NP_848519.1 | |
| XM_017014561.1 | 8 | 1533 | XP_016870050.1 | |
| XM_017014562.1 | 9 | 1612 | XP_016870051.1 | |
| XM_017014563.1 | 7 | 1410 | XP_016870052.1 | |
| XM_017014564.1 | 8 | 1534 | XP_016870053.1 | |
| AF035013.1 | 7 | 1428 | AAC98920.1 | |
| AF113130.1 | 5 | 2738 | AAF43778.1 | |
| AK075325.1 | 7 | 1317 | ||
| AK226135.1 | 2 | 4700 | ||
| AK294193.1 | 7 | 1486 | ||
| BC002655.2 | 7 | 1222 | AAH02655.1 | |
| BM982394.1 | ||||
| BX647489.1 | 5 | 2756 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035545 | Chr.9:87947580 - 88302726 on Build GRCh38 | Gain |
|
| nsv614788 | Chr.9:87801061 - 88227679 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map