Assay Details
Target Gene Details
Entrez Gene ID: | 79035 |
Gene Name: | nucleic acid binding protein 2 |
Gene Aliases: |
OBFC2B, SOSS-B1, SSB1, hSSB1 |
Location: |
Chr.12:56222039-56229858 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NABP2 | NM_024068.3 | 7 | 811 | NP_076973.1 |
| XM_005269146.2 | 7 | 822 | XP_005269203.1 | |
| XM_005269147.2 | 7 | 831 | XP_005269204.1 | |
| XM_005269148.4 | 7 | 806 | XP_005269205.1 | |
| XM_005269149.4 | 7 | 871 | XP_005269206.1 | |
| AY203942.1 | 7 | 770 | AAP34465.1 | |
| BC001079.2 | 6 | 821 | AAH01079.1 | |
| BC006171.1 | 7 | 785 | AAH06171.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
| nsv1051961 | Chr.12:55868436 - 56235643 on Build GRCh38 | Gain |
|
| nsv952186 | Chr.12:56224417 - 56245416 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map