Assay Details
Target Gene Details
Entrez Gene ID: | 140691 |
Gene Name: | tripartite motif containing 69 |
Gene Aliases: |
HSD-34, HSD34, RNF36, Trif |
Location: |
Chr.15:44736362-44767829 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIM69 | NM_001301144.1 | NP_001288073.1 | ||
| NM_001301145.1 | NP_001288074.1 | |||
| NM_001301146.1 | NP_001288075.1 | |||
| NM_080745.4 | NP_542783.2 | |||
| NM_182985.4 | NP_892030.3 | |||
| AF302088.1 | AAL55810.1 | |||
| AF302089.1 | AAL55811.1 | |||
| AK226115.1 | ||||
| AK292252.1 | ||||
| AL360161.1 | ||||
| AY305385.1 | AAQ75551.1 | |||
| BC024199.1 | AAH24199.1 | |||
| BC033314.1 | AAH33314.1 | |||
| BC047945.1 | AAH47945.1 | |||
| BI465168.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832990 | Chr.15:44698567 - 44870516 on Build GRCh38 | Gain |
|
| esv2760029 | Chr.15:44676670 - 45130644 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs138626529] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map