Assay Details
Target Gene Details
Entrez Gene ID: | 7126 |
Gene Name: | TNF alpha induced protein 1 |
Gene Aliases: |
B12, B61, BTBD34, EDP1, hBACURD2 |
Location: |
Chr.17:28335522-28347009 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFAIP1 | NM_021137.4 | 7 | 2052 | NP_066960.1 |
| XM_017024993.1 | 7 | 2130 | XP_016880482.1 | |
| AK300584.1 | ||||
| BC001643.2 | 7 | 1799 | AAH01643.1 | |
| BC001949.1 | 7 | 1799 | AAH01949.1 | |
| M80783.1 | 7 | 1756 | AAA58385.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2017 | Chr.17:28325611 - 28357716 on Build GRCh38 | Insertion |
|
| nsv519388 | Chr.17:28303453 - 28379927 on Build GRCh38 | Loss |
|
| nsv574653 | Chr.17:28227749 - 28501479 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map