Assay Details
Target Gene Details
Entrez Gene ID: | 26009 |
Gene Name: | zinc finger ZZ-type containing 3 |
Gene Aliases: |
ATAC1 |
Location: |
Chr.1:77561526-77683440 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZZZ3 | NM_001308237.1 | 14 | 2768 | NP_001295166.1 |
| NM_015534.5 | 15 | 4232 | NP_056349.1 | |
| XM_005270725.3 | 15 | 4307 | XP_005270782.1 | |
| XM_005270726.3 | 14 | 4208 | XP_005270783.1 | |
| XM_005270727.3 | 15 | 4304 | XP_005270784.1 | |
| XM_005270729.4 | 13 | 2718 | XP_005270786.1 | |
| XM_017000970.1 | 14 | 4205 | XP_016856459.1 | |
| XM_017000972.1 | 14 | 2736 | XP_016856461.1 | |
| AK001224.1 | 5 | 1605 | ||
| AK074119.1 | 10 | 4647 | BAB84945.1 | |
| AL080063.1 | 7 | 1796 | CAB45694.1 | |
| BC035079.2 | 14 | 2760 | AAH35079.1 | |
| BC035397.1 | 11 | 2900 | AAH35397.1 | |
| BC035818.2 | 11 | 3807 | AAH35818.1 | |
| BX640658.1 | 11 | 2320 | CAE45800.1 | |
| BX640766.1 | 11 | 3506 | CAE45870.1 | |
| BX641001.1 | 15 | 4233 | CAE46004.1 | |
| BX641096.1 | 15 | 4216 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1533 | Chr.1:77524819 - 77570594 on Build GRCh38 | Deletion |
|
| nsv546591 | Chr.1:77308728 - 77803522 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map