Assay Details
Target Gene Details
Entrez Gene ID: | 1576 |
Gene Name: | cytochrome P450 family 3 subfamily A member 4 |
Gene Aliases: |
CP33, CP34, CYP3A, CYP3A3, CYPIIIA3, CYPIIIA4, HLP, NF-25, P450C3, P450PCN1 |
Location: |
Chr.7:99756960-99784188 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP3A4 | NM_001202855.2 | 8 | 824 | NP_001189784.1 |
| NM_017460.5 | 8 | 827 | NP_059488.2 | |
| XM_017011780.1 | 8 | 825 | XP_016867269.1 | |
| AF182273.1 | 8 | 729 | AAF13598.1 | |
| AK298451.1 | 7 | 698 | ||
| AK298462.1 | 6 | 574 | ||
| AK312967.1 | 8 | 820 | ||
| AY606313.2 | 8 | 722 | ||
| BC069418.1 | 8 | 770 | AAH69418.1 | |
| BC101631.1 | 8 | 816 | AAI01632.1 | |
| D00003.1 | 8 | 780 | BAA00001.1 | |
| DQ924960.1 | 8 | 720 | ||
| J04449.1 | 8 | 785 | AAA35747.1 | |
| M13785.1 | 8 | 780 | AAA35742.1 | |
| M14096.1 | 8 | 788 | AAA35744.1 | |
| M18907.1 | 8 | 810 | AAA35745.1 | |
| X12387.1 | 8 | 789 | CAA30944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482109 | Chr.7:99756981 - 99784185 on Build GRCh38 | Gain |
|
| nsv970558 | Chr.7:99760896 - 99792284 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map