Assay Details
Target Gene Details
Entrez Gene ID: | 83666 |
Gene Name: | poly(ADP-ribose) polymerase family member 9 |
Gene Aliases: |
ARTD9, BAL, BAL1, MGC:7868 |
Location: |
Chr.3:122527910-122564676 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARP9 | NM_001146102.1 | 11 | 2896 | NP_001139574.1 |
| NM_001146103.1 | 11 | 2973 | NP_001139575.1 | |
| NM_001146104.1 | 11 | 2972 | NP_001139576.1 | |
| NM_001146105.1 | 11 | 2791 | NP_001139577.1 | |
| NM_031458.2 | 11 | 3078 | NP_113646.2 | |
| XM_005247820.2 | 11 | 3002 | XP_005247877.1 | |
| XM_017007304.1 | 11 | 2886 | XP_016862793.1 | |
| XM_017007305.1 | 11 | 2781 | XP_016862794.1 | |
| XM_017007306.1 | 11 | 2862 | XP_016862795.1 | |
| AF307338.1 | 11 | 2979 | AAK07558.1 | |
| AF307339.1 | 11 | 2874 | AAK07559.1 | |
| AK123412.1 | 4 | 2425 | ||
| AK130147.1 | 6 | 1427 | ||
| AK292959.1 | 11 | 2751 | ||
| AK299107.1 | 11 | 2865 | ||
| AL713679.1 | 4 | 1179 | CAD28483.1 | |
| AL832929.1 | 4 | 1111 | CAH56290.1 | |
| BC039580.1 | 10 | 2749 | AAH39580.1 | |
| BU675792.1 | 1 | 282 | ||
| BX648869.1 | 9 | 1618 | CAH56133.1 |
More Information
Set Membership: |
|

Genomic Map