Assay Details
Target Gene Details
Entrez Gene ID: | 51311 |
Gene Name: | toll like receptor 8 |
Gene Aliases: |
CD288 |
Location: |
Chr.X:12906620-12923169 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLR8 | NM_016610.3 | 3 | 3074 | NP_057694.2 |
| NM_138636.5 | 2 | 2937 | NP_619542.1 | |
| XM_011545529.1 | 3 | 3225 | XP_011543831.1 | |
| XM_011545530.2 | 3 | 3053 | XP_011543832.1 | |
| AB445666.1 | 1 | 2850 | ||
| AF245703.1 | 2 | 2898 | AAF78036.1 | |
| AF246971.1 | 3 | 3055 | AAF64061.1 | |
| AK075117.1 | 1 | 415 | BAC11412.1 | |
| AK297024.1 | 1 | 744 | ||
| AK307562.1 | 1 | 815 | ||
| AK314240.1 | 2 | 2937 | ||
| AY358296.1 | 2 | 2906 | AAQ88663.1 | |
| BC101074.1 | 2 | 2904 | AAI01075.1 | |
| BC101075.2 | 2 | 2904 | AAI01076.1 | |
| BC101076.2 | 2 | 2904 | AAI01077.1 | |
| BC101077.1 | 2 | 2904 | AAI01078.1 |
Target Gene Details
Entrez Gene ID: | 349408 |
Gene Name: | TLR8 antisense RNA 1 |
Gene Aliases: |
GS1-324M7.6 |
Location: |
Chr.X:12902817-12943300 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLR8-AS1 | NR_030727.1 | |||
| BI907428.1 |
More Information
Set Membership: |
|

Genomic Map