Assay Details
Target Gene Details
Entrez Gene ID: | 132321 |
Gene Name: | chromosome 4 open reading frame 33 |
Gene Aliases: |
- |
Location: |
Chr.4:129093674-129112688 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C4orf33 | NM_001099783.1 | 2 | 166 | NP_001093253.1 |
| NM_173487.2 | 2 | 438 | NP_775758.2 | |
| XM_005262734.4 | 2 | 433 | XP_005262791.1 | |
| XM_017007722.1 | 2 | 1495 | XP_016863211.1 | |
| XM_017007723.1 | 2 | 1245 | XP_016863212.1 | |
| XM_017007724.1 | 2 | 421 | XP_016863213.1 | |
| AK091022.1 | 2 | 427 | BAC03568.1 | |
| AK309229.1 | 2 | 412 | ||
| BC032582.1 | 2 | 369 | AAH32582.1 | |
| BE566349.1 | 2 | 121 | ||
| BP871612.1 | 2 | 438 | ||
| BQ549846.1 | 2 | 197 | ||
| BX538164.1 | 2 | 379 | CAD98044.1 | |
| CN483524.1 | 2 | 166 |
More Information
Set Membership: |
|

Genomic Map