Assay Details
Target Gene Details
Entrez Gene ID: | 199692 |
Gene Name: | zinc finger protein 627 |
Gene Aliases: |
- |
Location: |
Chr.19:11575255-11619159 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF627 | NM_001290083.1 | 5 | 1309 | NP_001277012.1 |
| NM_001290084.1 | 4 | 1143 | NP_001277013.1 | |
| NM_001290085.1 | 4 | 1150 | NP_001277014.1 | |
| NM_145295.3 | 4 | 1154 | NP_660338.1 | |
| XM_011527780.2 | 4 | 1042 | XP_011526082.1 | |
| AK074846.1 | 4 | 1062 | BAC11240.1 | |
| AK075063.1 | 1 | 242 | BAC11379.1 | |
| AK095639.1 | 4 | 1080 | ||
| BC006279.2 | 4 | 1075 | AAH06279.2 | |
| BC069232.1 | 4 | 1039 | ||
| BC098416.1 | 4 | 1125 | AAH98416.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509721 | Chr.19:11525216 - 11691390 on Build GRCh38 | Insertion |
|
| esv2763196 | Chr.19:11604699 - 11644922 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map