Assay Details
Target Gene Details
Entrez Gene ID: | 284 |
Gene Name: | angiopoietin 1 |
Gene Aliases: |
AGP1, AGPT, ANG1 |
Location: |
Chr.8:107249482-107498055 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGPT1 | NM_001146.4 | 9 | 2589 | NP_001137.2 |
| NM_001199859.2 | 9 | 2586 | NP_001186788.1 | |
| NM_001314051.1 | 8 | 1771 | NP_001300980.1 | |
| BC152411.1 | 9 | 2443 | ||
| BC152419.1 | 9 | 2455 | ||
| BX648814.1 | 9 | 2590 | CAI45984.1 | |
| D13628.2 | 9 | 2443 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2764100 | Chr.8:107208443 - 107457005 on Build GRCh38 | Gain |
|
| nsv611851 | Chr.8:107212459 - 107473684 on Build GRCh38 | Gain |
|
| nsv1028726 | Chr.8:106804613 - 107380319 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map