Assay Details
Target Gene Details
Entrez Gene ID: | 54676 |
Gene Name: | GTP binding protein 2 |
Gene Aliases: |
- |
Location: |
Chr.6:43620481-43629246 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTPBP2 | NM_001286216.1 | 12 | 2212 | NP_001273145.1 |
| NM_019096.4 | 12 | 2407 | NP_061969.3 | |
| AB024574.1 | 13 | 2250 | BAB12431.1 | |
| AF168990.1 | 12 | 2204 | AAF78884.1 | |
| AJ420518.1 | 7 | 1570 | ||
| AK000430.1 | BAA91160.1 | |||
| AL834331.1 | 12 | 2189 | CAD38999.1 | |
| BC020980.2 | 8 | 1823 | AAH20980.2 | |
| BC028347.2 | 10 | 2115 | AAH28347.2 | |
| BC064968.1 | 12 | 2360 | AAH64968.1 | |
| BU736981.1 | 1 | 637 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5281 | Chr.6:43611090 - 43655905 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76645046] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map