Assay Details
Target Gene Details
Entrez Gene ID: | 1134 |
Gene Name: | cholinergic receptor nicotinic alpha 1 subunit |
Gene Aliases: |
ACHRA, ACHRD, CHRNA, CMS1A, CMS1B, CMS2A, FCCMS, SCCMS |
Location: |
Chr.2:174747592-174764472 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNA1 | NM_000079.3 | 8 | 1234 | NP_000070.1 |
| NM_001039523.2 | 9 | 1309 | NP_001034612.1 | |
| XM_017003256.1 | 8 | 1479 | XP_016858745.1 | |
| XM_017003257.1 | 7 | 1401 | XP_016858746.1 | |
| AK291338.1 | 8 | 1175 | ||
| AK299445.1 | 9 | 1298 | ||
| AK315312.1 | 8 | 1234 | ||
| CD013889.1 | 9 | 1249 | ||
| S77094.1 | 8 | 1204 | AAD14247.1 | |
| Y00762.1 | 8 | 1204 | CAA68731.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv459948 | Chr.2:174710921 - 174762320 on Build GRCh38 | Loss |
|
| esv3575313 | Chr.2:174591458 - 175085724 on Build GRCh38 | Gain |
|
| nsv1012645 | Chr.2:174704308 - 174757118 on Build GRCh38 | Loss |
|
| esv3584180 | Chr.2:174741400 - 174771250 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map