Assay Details
Target Gene Details
Entrez Gene ID: | 4692 |
Gene Name: | necdin, MAGE family member |
Gene Aliases: |
HsT16328, PWCR |
Location: |
Chr.15:23685407-23687303 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDN | NM_002487.2 | 1 | 566 | NP_002478.1 |
| AK129654.1 | 2 | 464 | ||
| AK312779.1 | 1 | 561 | ||
| BC008750.1 | 1 | 543 | AAH08750.1 | |
| KJ534888.1 | 1 | 480 | ||
| KJ535010.1 | ||||
| KU178221.1 | 1 | 480 | ||
| KU178222.1 | 2 | 282 | ||
| KU178223.1 | ||||
| KU178224.1 | ||||
| KU178225.1 | ||||
| U35139.1 | 1 | 538 | AAB39469.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892632 | Chr.15:23375712 - 24546172 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map