Assay Details
Target Gene Details
Entrez Gene ID: | 1047 |
Gene Name: | calmegin |
Gene Aliases: |
- |
Location: |
Chr.4:140388453-140427661 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLGN | NM_001130675.1 | 16 | 2580 | NP_001124147.1 |
| NM_004362.2 | 15 | 2496 | NP_004353.1 | |
| D86322.1 | 15 | 2463 | BAA22590.1 |
More Information
Set Membership: |
|

Genomic Map